Learn More
12-Hydroxystearic acid, 95%, Thermo Scientific Chemicals
Description
12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 72°C to 76°C |
Color | White |
Linear Formula | CH3(CH2)5CH(OH)(CH2)10CO2H |
Quantity | 25 g |
Beilstein | 1726730 |
Solubility Information | Soluble in ether,alcohol and chloroform. Insoluble in water. |
Formula Weight | 300.48 |
Percent Purity | 95% |
Physical Form | Crystalline |
Chemical Name or Material | 12-Hydroxystearic acid |
Safety and Handling
EINECSNumber | 203-366-1 |
RTECSNumber | W13700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.