Alfa Aesar™ 12-Hydroxystearic acid, tech.

Neoplastic agent used in surface active and lubricating formulations

Manufacturer: Alfa Aesar™ 1526330

Quantity 250g

 View more versions of this product

Catalog No. AA1526330

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 106-14-9
    Formula Weight 288.54
    MDL Number MFCD00004592
    Synonym (+/-)-12-Hydroxyoctadecanoic acid
    Chemical Name or Material 12-Hydroxystearic acid
    Grade Technical Grade

    Specifications

    Formula Weight 288.54
    Physical Form Crystalline
    CAS 106-14-9
    TSCA TSCA
    MDL Number MFCD00004592
    Synonym (+/-)-12-Hydroxyoctadecanoic acid
    Quantity 250g
    Melting Point 80° to 81°C
    Chemical Name or Material 12-Hydroxystearic acid
    Grade Technical Grade
  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit