Alfa Aesar™ 12-Tricosanone, 97%

Antiblocking agent

Manufacturer: Alfa Aesar™ 4144422

Quantity 100g

 View more versions of this product

Catalog No. AA4144422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 540-09-0
    Formula Weight 338.62
    MDL Number MFCD00026562
    Synonym Diundecyl ketone
    Chemical Name or Material 12-Tricosanone

    Specifications

    Formula Weight 338.62
    MDL Number MFCD00026562
    RTECS Number YD0805000
    Physical Form Powder
    CAS 540-09-0
    Synonym Diundecyl ketone
    Melting Point 66° to 68°C
    TSCA TSCA
    Quantity 100g
    Chemical Name or Material 12-Tricosanone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit