Alfa Aesar™ 12-Tungstophosphate hydrate, Reagent Grade

Manufacturer: Alfa Aesar™ 1182718

Quantity 50g

 View more versions of this product

Catalog No. AA1182718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12501-23-4
    Formula Weight 2880.20 (Anhydrous)
    MDL Number MFCD00011342
    UN Number UN3260
    Synonym 12-Phosphotungstic acid hydrate
    Chemical Name or Material 12-Tungstophosphate hydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 2880.20 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    Physical Form Powder
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    MDL Number MFCD00011342
    UN Number UN3260
    Synonym 12-Phosphotungstic acid hydrate
    Color Off-white to pale yellow
    Quantity 50g
    Sensitivity Moisture & Light Sensitive
    CAS 12501-23-4
    Melting Point 95°C
    Chemical Name or Material 12-Tungstophosphate hydrate
    Grade Reagent Grade

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit