Learn More
Cole-Parmer Monarch DC6-1-12-0-0-0-3-0 12 universal inputs, 6 math channels, 100/240 Vac, 2GB internal memory  DFS Item
Manufacturer:  Cole-Parmer 4900633
The DataChart 6000 is like no other paperless recorder available! It has 6 or 12 universal inputs, up to 4 pulse/frequency inputs and 6 or 12 relay outputs. Networkable using standard Ethernet port; IP65/NEMA4 rated front panel. Touchscreen control.
Catalog No. 50-631-756