Learn More
Brady™ 120/277V Breaker Lockout Pouch with Brady Steel Padlocks & Tags
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ 120/277V Breaker Lockout Pouch is a convienent pouch contains all the devices you need to lockout most 120/277V breakers
Manufacturer:  Brady™ 99303
Catalog No. 19-104-190
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical risk lockouts
4.75 in.
2 in.
7.5 in.
Safety
Black on red
(1) lockout pouch; (2) single-pole circuit breakers, 277v; (2) no-hole circuit breakers; (2) multi-pole circuit breakers; (2) lockout cleats, 277v; (2) kd steel padlocks, 0.75 in. shackle, red; (2) lockout tags
Nylon
2 in. x 7.5 in. x 4.75 in.