Learn More
FUJIFILM Wako Endotoxin Indicator Vials
Ready-to-use challenge vials composed of E. coli O55:B5 (> 1000 EU/vial) for performing depyrogenation validation studies
Manufacturer:  FUJIFILM Wako EIV025
Catalog No. 12-333-028
Add to cart
Description
Description
  • E. coli O55:B5 (>1000 EU/vial)
  • Ready-to-use vials, no preparation needed
  • Used for validation studies
Specifications
Specifications
<1000 EU/vial
Reagent
Coating on inner surface of glass vial
25 vials with >1000 EU/vial
25 Vials