Learn More
Ly-6K Rat anti-Mouse, PE, Clone: B33fc8k, eBioscience™ Invitrogen™ Non-distribution product as customer accommodation.
Rat Monoclonal Antibody
Catalog No. 12-592-980
Add to cart
Description
Description
This B33fc8k monoclonal antibody reacts with mouse Ly-6k, a member of the Ly6/uPAR superfamily. Ly-6k is a GPI linked protein predominantly found on the cell surface. One study suggests expression may also be non-membranous. Expression is found on B cells and reported to be on plasma cells as well as cells in the testis. Using the B33fc8k monoclonal antibody, expression is on a B lymphocyte cell subset in the spleen that correlates to transitional cells (CD23-IgM-). In humans Ly-6k has been found associated with carcinomas of the head and testis. Applications Reported: This B33fc8k antibody has been reported for use in flow cytometric analysis. Applications Tested: This B33fc8k antibody has been tested by flow cytometric analyis of mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Ly-6K Monoclonal antibody specifically detects Ly-6K in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
Ly-6K
Flow Cytometry
B33fc8k
PE
LY6K
2410015A16Rik; 3110035B01Rik; cancer/testis antigen 97; CO16; CT97; FLJ35226; HSJ001348; Ly6k; Ly-6K; lymphocyte antigen 6 complex, locus K; lymphocyte antigen 6 complex, locus K-like; lymphocyte antigen 6K; mLy-6K; up-regulated in lung cancer 10; URLC10
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
76486
Liquid
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9CWP4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse