Learn More
Cole-Parmer Cedarberg 8550-255 Adjustable Safety Splash Shield, Polycarbonate, 12" W x 12" H  DFS Item
Manufacturer:  Cole-Parmer 4105203
Durable, clear polycarbonate shields are adjustable 360°>. Magnetic base holds fully flexible arm and shield firmly in place. Remove film on both sides to be clear.
Catalog No. NC9764259