Learn More
EARTHSAFE CHEMICAL ALTERNATIVES LLC 13.1G Sporicidal Tablet Cleaner And Disinfectant
Supplier:  EARTHSAFE CHEMICAL ALTERNATIVES LLC ESPO13.1GLP
Encompass_Preferred
13.1g Sporicidal NaDCC Cleaner/Disinfectant Tablet - Kill C Diff in 4 min / Kills Bacteria in Biofilm - LOOSE PACK (256ct tub / 2 tubs per Case). For use with EV110WR and EV160WR.
Catalog No. NC1912135
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
EARTHSAFE CHEMICAL ALTERNATIVES LLC 13.1G Sporicidal Tablet Cleaner And Disinfectant
Your feedback has been submitted: Thank you for helping us improve our website.