Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid, 98%, 90-100 μg/mL in ethanol

Click to view available options
Quantity:
0.5mg
100μg
29623-28-7
C18H32O3
296.451
MFCD00065833
HNICUWMFWZBIFP-IRQZEAMPSA-N
9z,11e,13s-13-hydroxyoctadeca-9,11-dienoic acid,13-hode,13 s-hydroxyoctadeca-9z,11e-dienoic acid,13 s-hode,13s-hydroxyoctadecadienoic acid,13s-hode,13-hydroxyoctadecadienoic acid,9z, 11e-13s-13-hydroxyoctadeca-9,11-dienoic acid,13s-hydroxy-9z,11e-octadecadienoic acid,13 s-hydroxy-9 z ,11 e-octadecadienoic acid
6443013
CHEBI:34154
(9Z,11E,13S)-13-hydroxyoctadeca-9,11-dienoic acid
CCCCCC(C=CC=CCCCCCCCC(=O)O)O
29623-28-7
C18H32O3
296.451
MFCD00065833
HNICUWMFWZBIFP-IRQZEAMPSA-N
9z,11e,13s-13-hydroxyoctadeca-9,11-dienoic acid,13-hode,13 s-hydroxyoctadeca-9z,11e-dienoic acid,13 s-hode,13s-hydroxyoctadecadienoic acid,13s-hode,13-hydroxyoctadecadienoic acid,9z, 11e-13s-13-hydroxyoctadeca-9,11-dienoic acid,13s-hydroxy-9z,11e-octadecadienoic acid,13 s-hydroxy-9 z ,11 e-octadecadienoic acid
6443013
CHEBI:34154
(9Z,11E,13S)-13-hydroxyoctadeca-9,11-dienoic acid
CCCCCC(C=CC=CCCCCCCCC(=O)O)O
Quantity 0.5mg
UN Number UN1170
Formula Weight 296.45
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid, 90 to 100μg/mL in ethanol

GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHANOL SOLUTIONS

storageNote1 : -30° to -10°C