Learn More
Sino Biological Human RSV (B1) glycoprotein G / RSV-G Protein (His Tag) 100ug
Supplier:  Sino Biological 13029V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human RSV strain B1 glycoprotein G (O36633-1) (His 67-Ala 299) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-8180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.