Learn More
Epredia™ Lab Vision™ 14.3.3σ Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Take advantage of a quality product that delivers accurate results with the Epredia™ 14.3.3σ Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1185P1ABX
Description
14.3.3 proteins are a family of small, widely expressed, highly conserved cytosolic proteins. 14.3.3 proteins bind to and influence the activities of a diverse group of molecules involved in signal transduction, cell cycle regulation and apoptosis, including Raf, PKC, Bad, Cbl, and c-Bcr. Interactions between 14.3.3 and target proteins are strongly influenced by the phosphorylation state of 14.3.3 and the target protein. 14.3.3 σ shares cyclin-Cdk2 binding motifs with different cell cycle regulators, including p107, p130, p21Cip1, p27Kip1, and p57Kip2 and is associated with cyclin-Cdk complexes in vitro and in vivo.Host Species: Mouse
Clone: 1433S01
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human 14.3.3σ protein
Molecular Weight: 26-30kDa
Positive Control: HT29 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
14.3.3sigma Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Signal Transduction | |
Human | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1433S01 | |
Unconjugated | |
Recombinant human 1433 sigma protein | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.