Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ 14.3.3σ Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ 14.3.3σ Ab-1, Mouse Monoclonal Antibody.

$156.00 - $1001.00

Specifications

Antigen 14.3.3sigma Ab-1
Clone 1433S01
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1185P
Accessories View Documents
Thermo Scientific™
MS1185P
Purified Ab with BSA and Azide 1mL
Each for $1,001.00
Add to cart
 
MS1185P0
Accessories View Documents
Thermo Scientific™
MS1185P0
Purified Ab with BSA and Azide 100μL
Each for $156.00
Add to cart
 
MS1185P1
Accessories View Documents
Thermo Scientific™
MS1185P1
Purified Ab with BSA and Azide 500μL
Each for $483.00
Add to cart
 
MS1185P1ABX
Accessories View Documents
Thermo Scientific™
MS1185P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $527.50
Add to cart
 
MS1185PABX
Accessories View Documents
Thermo Scientific™
MS1185PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $869.00
Add to cart
 
MS1185R7
Accessories View Documents
Thermo Scientific™
MS1185R7
Ready-to-use for IHC (UVLP) 7mL
Each for $417.00
Add to cart
 
MS1185R1
View Documents
Thermo Scientific™
MS1185R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ 14.3.3σ Ab-1, Mouse Monoclonal Antibody.

14.3.3 proteins are a family of small, widely expressed, highly conserved cytosolic proteins. 14.3.3 proteins bind to and influence the activities of a diverse group of molecules involved in signal transduction, cell cycle regulation and apoptosis, including Raf, PKC, Bad, Cbl, and c-Bcr. Interactions between 14.3.3 and target proteins are strongly influenced by the phosphorylation state of 14.3.3 and the target protein. 14.3.3 σ shares cyclin-Cdk2 binding motifs with different cell cycle regulators, including p107, p130, p21Cip1, p27Kip1, and p57Kip2 and is associated with cyclin-Cdk complexes in vitro and in vivo.

Host Species: Mouse

Clone: 1433S01

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant human 14.3.3σ protein

Molecular Weight: 26-30kDa

Positive Control: HT29 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

14.3.3sigma Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Primary
IgG2a kappa
1433S01
Monoclonal
Murine
Signal Transduction
Recombinant human 1433 sigma protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit