Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 beta/epsilon/zeta Mouse anti-Human, Mouse, Rat, Clone: 3C8, Novus Biologicals™
Mouse Monoclonal Antibody
$218.00 - $521.00
Specifications
Antigen | 14-3-3 beta/epsilon/zeta |
---|---|
Clone | 3C8 |
Dilution | Western Blot 1-10 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Description
14-3-3 beta/epsilon/zeta Monoclonal specifically detects 14-3-3 beta/epsilon/zeta in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
14-3-3 beta/epsilon/zeta | |
Western Blot 1-10 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Recombinant human 14-3-3 beta. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
3C8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Rat, Human | |
IgG2b | |
Protein A purified | |
Recognizes Human, Mouse and Rat 14-3-3 beta/epsilon/zeta. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title