Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 beta/epsilon/zeta, Mouse anti-Human,Mouse,Rat, DyLight 680, Clone: 3C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP197885FR

Catalog No. NB004744


Add to cart

Description

Description

14-3-3 beta/epsilon/zeta Monoclonal specifically detects 14-3-3 beta/epsilon/zeta in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

14-3-3 beta/epsilon/zeta
3C8
Western Blot
Purified, Conjugated
Recombinant human 14-3-3 beta.
Protein A purified
RUO
Primary
Human, Mouse, Rat
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit