Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 epsilon, Mouse anti-Human, DyLight 594, Clone: CPTC-YWHAE-1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279944DL594
Description
14-3-3 epsilon Monoclonal antibody specifically detects 14-3-3 epsilon in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
14-3-3 epsilon | |
CPTC-YWHAE-1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
YWHAE | |
Recombinant human full-length 14-3-3 epsilon protein | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide | |
Mouse | |
IgG2a Kappa | |
0.1 mL | |
Cancer, Lipid and Metabolism | |
Primary | |
7531 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title