Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 epsilon Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299633JF549

Catalog No. NB062124


Add to cart

Description

Description

14-3-3 epsilon Polyclonal antibody specifically detects 14-3-3 epsilon in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

14-3-3 epsilon
Janelia Fluor 549
50mM Sodium Borate
14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide
Produced in rabbits immunized with purified, recombinant 14-3-3 epsilon (NP_006752.1; Met1-Gln255).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Immunofluorescence
Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Cancer, Lipid and Metabolism
Primary
7531
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit