Learn More
14-3-3 epsilon Recombinant Rabbit Monoclonal Antibody (5H10L5), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific 702241
Description
This antibody is predicted to react with Monkey, Sheep, Rat and Mouse. Recombit rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombit rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
14-3-3 epsilon belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the mouse ortholog. It interacts with CDC25 phosphatases, RAF1 and IRS1 proteins, suggesting its role in diverse biochemical activities related to signal transduction, such as cell division and regulation of insulin sensitivity. It has also been implicated in the pathogenesis of small cell lung cancer. Two transcript variants, one protein-coding and the other non-protein-coding, have been found for this gene.Specifications
14-3-3 epsilon | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P62258, P62259, P62260 | |
YWHAE | |
Peptides corresponding to Human YWHAE (aa 70-87, 238-255) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
5H10L5 | |
Unconjugated | |
YWHAE | |
14-3-3 protein epsilon | |
Rabbit | |
Protein A | |
RUO | |
22627, 29753, 7531 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Research Use Only