Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 Rabbit anti-Human, DyLight 594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299637DL594

Catalog No. NB062154


Add to cart

Description

Description

14-3-3 Polyclonal specifically detects 14-3-3 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

14-3-3
DyLight 594
50mM Sodium Borate
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Produced in rabbits immunized with purified, recombinant 14-3-3 (P27348; Met1-Asn245).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction
Primary
10971
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit