Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 sigma/Stratifin Mouse anti-Human, Clone: CPTC-SFN-2, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | 14-3-3 sigma/Stratifin |
---|---|
Clone | CPTC-SFN-2 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122126
|
Novus Biologicals
NBP30732620UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
14-3-3 sigma/Stratifin Monoclonal specifically detects 14-3-3 sigma/Stratifin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
14-3-3 sigma/Stratifin | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Phospho Specific | |
10 mM PBS with 0.05% BSA | |
2810 | |
IgG1 κ | |
Protein A or G purified |
CPTC-SFN-2 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS | |
Recombinant human full-length 14-3-3 sigma/Stratifin protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title