Learn More
Cayman ChemicalSupplier Diversity Partner 14(S)-HDHA, 119433-37-3, 100 µg  DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 15253100
An oxygenation product of DHA that serves as a precursor to the anti-inflammatory, pro-resolving maresin 1; doses as low as 0.2 ng/mouse resulted in reduced infiltration of PMNs into sites of inflammation
Catalog No. NC1481545