Learn More
Keystone Cleanroom Products 1422A Tyvek Autoclavable Bowl Cover, with Steam Indicator
SDP
Keystone Cleanroom Products 1422A Tyvek™ Autoclavable Bowl Covers prevent microbial and particulate contamination to critical parts
$1474.00 - $6447.00
Specifications
Autoclavable Autoclavable
Color White
Material Tyvek
View More Specs
Products 15
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Price Quantity & Availability  
19-990-002
SDP
View Documents
Keystone Cleanroom Products
BWLT2A20
20 in. 50.8 cm
Case of 500 for $4,009.00
Add to Cart
19-990-003
SDP
View Documents
Keystone Cleanroom Products
BWLT2A24
24 in. 61 cm
Case of 500 for $4,508.00
Add to Cart
19-990-004
SDP
View Documents
Keystone Cleanroom Products
BWLT2A26
26 in. 66 cm
Case of 500 for $5,182.00
Add to Cart
19-990-005
SDP
View Documents
Keystone Cleanroom Products
BWLT2A30
30 in. 76.2 cm
Case of 500 for $6,447.00
Add to Cart
19-990-006
SDP
View Documents
Keystone Cleanroom Products
BWLT2A36
36 in. 91.4 cm
Case of 100 for $1,474.00
Add to Cart
19-990-007
SDP
View Documents
Keystone Cleanroom Products
BWLT2A38
38 in. 96.5 cm
Case of 100 for $2,754.00
Add to Cart
19-990-008
SDP
View Documents
Keystone Cleanroom Products
BWLT2A42
42 in. 106.7 cm
Case of 50 for $2,160.00
Add to Cart
19-990-009
SDP
View Documents
Keystone Cleanroom Products
BWLT2A46
46 in. 116.8 cm
Case of 50 for $2,285.00
Add to Cart
19-990-010
SDP
View Documents
Keystone Cleanroom Products
BWLT2A48
48 in. 122 cm
Case of 50 for $2,775.00
Add to Cart
19-990-012
SDP
View Documents
Keystone Cleanroom Products
BWLT2A54
54 in. 137.2 cm
Case of 50 for $3,225.00
Add to Cart
19-990-013
SDP
View Documents
Keystone Cleanroom Products
BWLT2A56
56 in. 142.2 cm
Case of 50 for $3,254.00
Add to Cart
19-990-014
SDP
View Documents
Keystone Cleanroom Products
BWLT2A60
60 in. 152.4 cm
Case of 50 for $3,590.00
Add to Cart
17-005-934
SDP
View Documents
Keystone Cleanroom Products
BWLT2A72
72 in. 183 cm
Case of 50 for $3,448.00
Add to Cart
17-005-935
SDP
View Documents
Keystone Cleanroom Products
BWLT2A90
90 in. 229 cm
Case of 50 for $3,978.00
Add to Cart
17-010-183
SDP
View Documents
Keystone Cleanroom Products
BWLT2A44
44 in. 112 cm
Case of 50 for $1,995.00
Add to Cart
Description
Description
  • Prevent microbial and particulate contamination to critical parts, while the fill line set-up activities are performed
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam Sterilization Indicator - Autoclavable at 121°C to 125°C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Specifications
Specifications
Autoclavable
White
Tyvek