Learn More
Keystone Cleanroom Products 1422A Tyvek™ Autoclave Bags
1422A Tyvek™ Autoclave Bags with drawstring closure create a superior microbial barrier
$1047.00 - $3340.00
Specifications
Description 1422A Tyvek™
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type String closure
Color White
View More Specs
Products 15
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-990-105
View Documents
Keystone Cleanroom Products
BGT2A0314D
(3 in. x 14 in.)
Case of 250 for $3,340.00
Add to cart
19-042-730
View Documents
Keystone Cleanroom Products
BGT2A0212
2 x 12 in.
Case of 500 for $1,472.00
Add to cart
19-990-106
View Documents
Keystone Cleanroom Products
BGT2A1526D
(15 in. x 26 in.)
Case of 250 for $2,750.00
Add to cart
19-042-731
View Documents
Keystone Cleanroom Products
BGT2A0512D
5 x 12 in.
Case of 250 for $1,134.00
Add to cart
19-042-733
View Documents
Keystone Cleanroom Products
BGT2A0515D
5 x 15 in.
Case of 250 for $1,204.00
Add to cart
19-042-734
View Documents
Keystone Cleanroom Products
BGT2A1007D
10 x 7 in.
Case of 250 for $1,047.00
Add to cart
19-042-735
View Documents
Keystone Cleanroom Products
BGT2A1026D
10 x 26 in.
Case of 250 for $1,542.00
Add to cart
19-042-736
View Documents
Keystone Cleanroom Products
BGT2A1062
10 x 62 in.
Case of 250 for $1,843.00
Add to cart
19-042-737
View Documents
Keystone Cleanroom Products
BGT2A1246
12 x 46 in.
Case of 250 for $1,709.00
Add to cart
19-042-738
View Documents
Keystone Cleanroom Products
BGT2A1622D
16 x 22 in.
Case of 250 for $1,819.00
Add to cart
19-042-739
View Documents
Keystone Cleanroom Products
BGT2A2432D
24 x 32 in.
Case of 250 for $2,220.00
Add to cart
19-042-740
View Documents
Keystone Cleanroom Products
BGT2A2462
24 x 62 in.
Case of 250 for $2,950.00
Add to cart
19-042-741
View Documents
Keystone Cleanroom Products
BGT2A3036
30 x 36 in.
Case of 250 for $2,110.00
Add to cart
19-042-743
View Documents
Keystone Cleanroom Products
BGT2A4242
42 x 42 in.
Case of 125 for $1,330.00
Add to cart
19-042-744
View Documents
Keystone Cleanroom Products
BGT2A5054
50 x 54 in.
Case of 100 for $1,353.00
Add to cart
Description
Description
  • Manufactured and packaged in a controlled cleanroom environment
  • Designed for steam sterilization of large, or oddly shaped pharmaceutical equipment
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Closed via the adjustable Tyvek™ drawstring closure system
  • Eliminates the need to cut material or removal of particulate generating autoclave tape
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
Specifications
Specifications
1422A Tyvek™
Certificate of Conformance (COC)
White
HDPE
Autoclavable
String closure
Steam Sterilization
250/Pk.