Learn More
Thermo Scientific 146iq-AABACN Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQAABACN
146iq Multigas Calibrator ∙Power Cord: 100-120 VAC, 50/60 Hz (IEC-A electrical plug; NA) ∙Communications: Serial RS232/RS485 ∙Calibration Sources: Gas Phase Titration and Photometer ∙Internal Zero Mass Flow Controller: 5 SLPM ∙Span Gas Mass Flow Controller #1: 200 SCCM ∙Span Gas Mass Flow Controller #2: None
Catalog No. 50-146-9079