Learn More
Thermo Scientific 146iq-AANBNC Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQAANBNC
146iq Multigas Calibrator ∙Power Cord: 100-120 VAC, 50/60 Hz (IEC-A electrical plug; NA) ∙Communications: Serial RS232/RS485 ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 10 SLPM ∙Span Gas Mass Flow Controller #1: None ∙Span Gas Mass Flow Controller #2: 5 LPM
Catalog No. 50-146-8995
Please call customer service for current pricing. 1-800-766-7000