Learn More
Thermo Scientific 146iq-ABACAA Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQABACAA
146iq Multigas Calibrator ∙Power Cord: 100-120 VAC, 50/60 Hz (IEC-A electrical plug; NA) ∙Communications: Analog and Digital ∙Calibration Sources: Gas Phase Titration ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 50 SCCM ∙Span Gas Mass Flow Controller #2: 1 LPM
Catalog No. 50-146-9192