Learn More
Thermo Scientific 146iq-ABNCCN Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQABNCCN
146iq Multigas Calibrator ∙Power Cord: 100-120 VAC, 50/60 Hz (IEC-A electrical plug; NA) ∙Communications: Analog and Digital ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 200 SCCM ∙Span Gas Mass Flow Controller #2: None
Catalog No. 50-146-9154
Please call customer service for current pricing. 1-800-766-7000