Learn More
Thermo Scientific 146iq-ACNCBA Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQACNCBA
146iq Multigas Calibrator ∙Power Cord: 100-120 VAC, 50/60 Hz (IEC-A electrical plug; NA) ∙Communications: Serial, Analog, and Digital ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 100 SCCM ∙Span Gas Mass Flow Controller #2: 1 LPM
Catalog No. 50-146-9286