Learn More
Thermo Scientific 146iq-BANCBN Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBANCBN
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: Serial RS232/RS485 ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 100 SCCM ∙Span Gas Mass Flow Controller #2: None
Catalog No. 50-146-9555