Learn More
Thermo Scientific 146iq-BBBCBN Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBBBCBN
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: Analog and Digital ∙Calibration Sources: Gas Phase Titration and Photometer ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 100 SCCM ∙Span Gas Mass Flow Controller #2: None
Catalog No. 50-146-9780