Learn More
Thermo Scientific 146iq-BCBCCA Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBCBCCA
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: Serial, Analog, and Digital ∙Calibration Sources: Gas Phase Titration and Photometer ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 200 SCCM ∙Span Gas Mass Flow Controller #2: 1 LPM
Catalog No. 50-146-9920