Learn More
Thermo Scientific 146iq-BCBCNA Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBCBCNA
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: Serial, Analog, and Digital ∙Calibration Sources: Gas Phase Titration and Photometer ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: None ∙Span Gas Mass Flow Controller #2: 1 LPM
Catalog No. 50-146-9908