Learn More
Thermo Scientific 146iq-BNAAAB Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBNAAAB
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: No I/O ∙Calibration Sources: Gas Phase Titration ∙Internal Zero Mass Flow Controller: 5 SLPM ∙Span Gas Mass Flow Controller #1: 50 SCCM ∙Span Gas Mass Flow Controller #2: 2 LPM
Catalog No. 50-146-9433