Learn More
Thermo Scientific 146iq-BNNANA Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBNNANA
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: No I/O ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 5 SLPM ∙Span Gas Mass Flow Controller #1: None ∙Span Gas Mass Flow Controller #2: 1 LPM
Catalog No. 50-146-9383