Learn More
Thermo Scientific 146iq-BNNCAN Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQBNNCAN
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-I electrical plug; CHN) ∙Communications: No I/O ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 50 SCCM ∙Span Gas Mass Flow Controller #2: None
Catalog No. 50-146-9416