Learn More
Thermo Scientific 146iq-CAABBB Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQCAABBB
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-C electrical plug; EU) ∙Communications: Serial RS232/RS485 ∙Calibration Sources: Gas Phase Titration ∙Internal Zero Mass Flow Controller: 10 SLPM ∙Span Gas Mass Flow Controller #1: 100 SCCM ∙Span Gas Mass Flow Controller #2: 2 LPM
Catalog No. 50-147-0127