Learn More
Thermo Scientific 146iq-CBBACB Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQCBBACB
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-C electrical plug; EU) ∙Communications: Analog and Digital ∙Calibration Sources: Gas Phase Titration and Photometer ∙Internal Zero Mass Flow Controller: 5 SLPM ∙Span Gas Mass Flow Controller #1: 200 SCCM ∙Span Gas Mass Flow Controller #2: 2 LPM
Catalog No. 50-147-0296