Learn More
Thermo Scientific 146iq-CBNCAN Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQCBNCAN
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-C electrical plug; EU) ∙Communications: Analog and Digital ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 20 SLPM ∙Span Gas Mass Flow Controller #1: 50 SCCM ∙Span Gas Mass Flow Controller #2: None
Catalog No. 50-147-0226