Learn More
Thermo Scientific 146iq-CCNBNB Multigas Calibrator DFS Item
Manufacturer:  Thermo Scientific 146IQCCNBNB
146iq Multigas Calibrator ∙Power Cord: 220 VAC, 50/60 Hz (IEC-C electrical plug; EU) ∙Communications: Serial, Analog, and Digital ∙Calibration Sources: No Optional Calibration Sources ∙Internal Zero Mass Flow Controller: 10 SLPM ∙Span Gas Mass Flow Controller #1: None ∙Span Gas Mass Flow Controller #2: 2 LPM
Catalog No. 50-147-0344