Learn More
Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors Available on GSA/VA Contract for Federal Government customers only.
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
Manufacturer:  Justrite™ 8825172
 View more versions of this product
Catalog No. 19-822-353
Add to cart
Description
Description
  • By joining centers together with these clips or the patented Sump-To-Sump Drain kit for a total of 66-gallon sump capacity, you will meet EPA and UFC requirements.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15 Gallon
Left-hinged, Manual-close
Light Neutral