Learn More
Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Self-Closing Doors
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Self-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
Supplier:  Justrite™ 8825272
 View more versions of this product
Catalog No. 19-822-362
Add to Cart
Description
Description
  • Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
  • Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
Specifications
Specifications
Right Door
Self-close
15 gal.
Light Neutral
Product Suggestions
Product Suggestions