Learn More
Cayman Chemical 15-keto ProstaglndIn F2a 1mg
SDP
Supplier:  Cayman Chemical 167201 MG
Encompass_Preferred
First metabolite of PGF2α in the 15-hydroxy PGDH pathway; a critical component in the goldfish and Atlantic salmon postovulatory pheromone
Catalog No. 50-296-8482