Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-PGDH/HPGD Mouse anti-Human, PE/Atto594, Clone: OTI2C10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270933PEATT594

Catalog No. N270933P594


Add to cart

Description

Description

15-PGDH/HPGD Monoclonal antibody specifically detects 15-PGDH/HPGD in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

15-PGDH/HPGD
OTI2C10
Flow Cytometry
Affinity Purified, Conjugated
HPGD
Full length recombinant protein of human 15-PGDH/HPGD (NP_000851) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Immunology, Innate Immunity
Primary
3248.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit