Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ 15-PGDH/HPGD Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer: Novus Biologicals™ NBP2796155PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human 15-PGDH/HPGD.Specifications
Sandwich ELISA | |
31.25-2000 pg/mL | |
Sandwich ELISA | |
3248 | |
Rabbit | |
5 Plates | |
Cancer, Immunology, Innate Immunity | |
Storage is content dependent. | |
Human |
31.25 pg/mL | |
HRP | |
15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
HPGD | |
Conjugated | |
Antibody Pairs | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human 15-PGDH/HPGD. | |
15-PGDH/HPGD | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title