Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-PGDH Mouse anti-Human, Clone: OTI2C10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525457

Catalog No. PIMA525457


Add to cart

Description

Description

This gene encodes a member of the short-chain nonmetalloenzyme alcohol dehydrogenase protein family. The encoded enzyme is responsible for the metabolism of prostaglandins, which function in a variety of physiologic and cellular processes such as inflammation. Mutations in this gene result in primary autosomal recessive hypertrophic osteoarthropathy and cranioosteoarthropathy. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

15-PGDH
Monoclonal
0.9 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P15428
15-PGDH, PGDH, PGDH1, PHOAR1, SDR36C1, 15-hydroxyprostaglandin dehydrogenase [NAD+], NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, prostaglandin dehydrogenase 1, short chain dehydrogenase/reductase family 36C, member 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3248
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2C10
Unconjugated
HPGD
Liquid
HPGD
Full length human recombit protein of HPGD produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit