Learn More
Thermo Scientific™ Orion™ AutoTration™ 500 Autosampler Accessories
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler
Manufacturer:  Thermo Scientific™ ASB150
 View more versions of this product
Catalog No. 13-641-965
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Maintain automated sample analysis with accessories for the legacy Thermo Scientific™ Orion™ AutoTration 500 Autosampler.
  • Interchangeable racks to accommodate sample size and volume
  • Choice of 48- and 28-position racks
  • Beaker sizes of 45-, 50- and 150-mL
  • Replacement electrode holders for each model
Description & Specifications
Specifications
AT5150 AutoTration-500 autosampler with 28-position rack
Autosampler 150 mL plastic beakers, 50 pack
AT5050 AutoTration-500 Autosampler with 48-position rack
Titrator Accessories
150 mL beakers
Autosampler 150 mL plastic beakers, 50 pack
50 beakers (150 mL)