Learn More
Mystaire™ Isola™ Filtered Workstation Pre-filter
SDP
For use with Isola™ filtered workstations. Protects the adsorption filter from submicron particulate.
Supplier:  Mystaire™ MYPRE20
Catalog No. 15-338-955
Add to Cart
Description
Description
  • Electrostatically charged fiber filter
  • Designed to protect adsorption filter from contaminants such as smoke, acid mists, and other submicron particulates
Specifications
Specifications
Particulate
1 in.
20 in.
457.2 x 508 x 25.4 mm (18 x 20 x 1 in.)
508 mm
Electrostatically Charged Fiber
Isola Filtered Workstation (48 in.)
18 in.
25.4 cm
457.2 mm
Prefilter
Product Suggestions
Product Suggestions