Learn More
Mystaire™ Latitude™ Fentanyl Filtered Hood
Safe, effective containment of fentanyl and its analogues during manipulation and laboratory analysis.
Supplier: Mystaire™ MYLFH72
Description
- Horziontal laminar airflow pattern
- EverSafe automatic safety controller
- Saf-T-Zone filtration system that allows for filter replacement while the hood is running and providing maximum containment
- Permanent safety HEPA filter
- Dark Blue base for contrast in powder clean up
- Waste port to attach Waste bag for disposal of packaging or gloves
Specifications
Filtered Hood | |
Thermally Fused Polypropylene | |
762 x 1829 x 1041 mm (30 x 72 x 41 in.) | |
1041 mm | |
762 mm |
Redundant HEPA and Bonded Carbon | |
579 x 1803 x 635 mm (22.8 x 71 x 25 in.) | |
41 in. | |
30 in. |