Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 1536-Well Polystyrene HiBase Non-binding Flat Bottom Microplate

Free of detectable DNase/RNase, human DNA and pyrogens.

$1802.00 - $1929.00

Specifications

Temperature Range (Metric) -20°C to +60°C
Max. RCF 1500 x g (swinging-bucket rotor)
Sterility Non-sterile
Bottom Flat
Well Shape Rounded Square
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07-000-910
View Documents
Greiner Bio-One
782900
Black
Case of 60 for $1,802.00
Add to cart
 
07-000-911
View Documents
Greiner Bio-One
782904
White
Case of 60 for $1,929.00
Add to cart
 
Description

Description

  • HiBase
  • Solid flat bottom
  • Rounded square well design
  • Alphanumeric well coding
  • Protein-repellent, non-binding treatment
  • Sterile

Service Plan

No

Specifications

Specifications

-20°C to +60°C
Non-sterile
Rounded Square
Yes
1536
6 to 12 μL
Yes
Not Autoclavable
1500 x g (swinging-bucket rotor)
Flat
Polystyrene
60
Protein-Repellent
Standard
15
Safety and Handling

Safety and Handling

ShelfLife : 4 Years after month of production

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit